Please login to your online account to display your discounted pricing

anti-Lipase A, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP154155

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP154155

  • / Each
 

Description & Specifications

Specifications

Antigen Lipase A
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blot
Concentration 0.67mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Affinity Purified
Formulation PBS, 0.1% BSA, 50% glycerol
Gene Accession No. P38571
Host Species Rabbit
Immunogen A genomic peptide made to an internal region of the human Lipase A protein (within residues 150-300). [Swiss-Prot P38571]
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 3988
Test Specificity Human and mouse. Immunogen sequence has 82% identity to pig and 73% identity to rat and cow.
Target Species Human
Target Species Murine