missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Lipase A, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A genomic peptide made to an internal region of the human Lipase A protein (within residues 150-300). [Swiss-Prot P38571]
Antigen Lipase A
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP154155 Novus Biologicals Canada
NBP154155
0.1mL Each for $329.00
NBP154155SS Novus Biologicals Canada
NBP154155SS
0.025mL Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
Lipase A
Affinity Purified
RUO
Human and mouse. Immunogen sequence has 82% identity to pig and 73% identity to rat and cow.
3988
0.67mg/mL
A genomic peptide made to an internal region of the human Lipase A protein (within residues 150-300). [Swiss-Prot P38571]
Polyclonal
Primary
Immunocytochemistry,Immunofluorescence,Immunohistochemistry,Immunohistochemistry (Paraffin),Simple Western,Western Blot
P38571
PBS, 0.1% BSA, 50% glycerol