Please login to your online account to display your discounted pricing

anti-LIN-28A, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224670

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224670

  • / Each
 

Description & Specifications

Specifications

Antigen LIN-28A
Format Purified
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Quantity 0.1mg
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 79727
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human isoform CRA_a and chimpanzee, 96% in dog, 92% in mouse and rat, and 88% homologous in cow ROR gamma. This antibody will detect ROR gamma and may detect ROR gamma (t), but can not be used to distinguish between the two proteins.
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Formulation 100μg in 200μL PBS containing 0.05% BSA
Gene Accession No. Q9H9Z2
Primary or Secondary Primary
Concentration 0.5mg/mL
Host Species Rabbit
Immunogen A portion of amino acids 100-150 of human LIN28 was used as the immunogen.
Regulatory Status RUO