Please login to your online account to display your discounted pricing

anti-LIN-28A, Polyclonal, Novus Biologicals™

$109.00 - $307.00

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 100-150 of human LIN28 was used as the immunogen.
Antigen LIN-28A
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224670

 
novus biologicals canada
NBP224670
Unlabeled 0.1mg Each for $307.00

NBP224670SS

 
novus biologicals canada
NBP224670SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 100-150 of human LIN28 was used as the immunogen.
Antigen LIN-28A
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human isoform CRA_a and chimpanzee, 96% in dog, 92% in mouse and rat, and 88% homologous in cow ROR gamma. This antibody will detect ROR gamma and may detect ROR gamma (t), but can not be used to distinguish between the two proteins.
Gene Accession No. Q9H9Z2
Gene ID (Entrez) 79727
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL