missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-LIM1, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen 175-162
Antigen LIM1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB10056626 Novus Biologicals Canada
NB10056626
0.2mL Each for $255.80
NB10056626T Novus Biologicals Canada
NB10056626SS
0.05mL Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
LIM1
Purified
RUO
In human muscle, a 60 kDa band is observed.
3975
0.5mg/mL
175-162
Polyclonal
Primary
Western Blot
NP_005559
PBS