Please login to your online account to display your discounted pricing

anti-Lgr5/GPR49, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP128904SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP128904SS

  • / Each
 

Description & Specifications

Specifications

Concentration 1.19mg/mL
Formulation PBS, 30% glycerol
Conjugate Unlabeled
Gene Accession No. O75473
Gene ID (Entrez) 8549
Host Species Rabbit
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Quantity 0.025mL
Cross Reactivity Human
Cross Reactivity Porcine
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Regulatory Status RUO
Immunogen Synthetic peptide made to an internal portion of the human GPR49/LGR5 protein (within residues 650-700). [Swiss-Prot No. O75473]
Antigen Lgr5/GPR49
Format Affinity Purified