Please login to your online account to display your discounted pricing

anti-Lgr5/GPR49, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen Synthetic peptide [KLH conjugated] made to the 2nd cytoplasmic loop of human GPR49/LGR5. [UniProt No. O75473]
Antigen Lgr5/GPR49
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NLS1235SS

 
novus biologicals canada
NLS1235SS
Unlabeled 0.025mL Each for $109.00

NLS1235

 
novus biologicals canada
NLS1235
Unlabeled 0.05mL Each for $309.05
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide [KLH conjugated] made to the 2nd cytoplasmic loop of human GPR49/LGR5. [UniProt No. O75473]
Antigen Lgr5/GPR49
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Test Specificity Human. Immunogen sequence has 81% identity with mouse.
Gene Accession No. O75473
Gene ID (Entrez) 8549
Formulation PBS, pH 7.4
Concentration 1mg/mL