Please login to your online account to display your discounted pricing

anti-Lgr5/GPR49, Polyclonal, Novus Biologicals™

$144.63 - $384.00

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the human GPR49/LGR5 protein (within residues 650-700). [Swiss-Prot No. O75473]
Antigen Lgr5/GPR49
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP128904SS

 
novus biologicals canada
NBP128904SS
Unlabeled 0.025mL Each for $144.63

NBP128904

 
novus biologicals canada
NBP128904
Unlabeled 0.1mL Each for $384.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the human GPR49/LGR5 protein (within residues 650-700). [Swiss-Prot No. O75473]
Antigen Lgr5/GPR49
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Gene Accession No. O75473
Gene ID (Entrez) 8549
Formulation PBS, 30% glycerol
Concentration 1.19mg/mL