Please login to your online account to display your discounted pricing

anti-Leukotriene B4 R1, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1-60 from human BLT1 was used as the immunogen for this antibody (1st extracellular domain).
Antigen Leukotriene B4 R1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227422

 
novus biologicals canada
NBP227422
Unlabeled 0.1mg Each for $255.80

NBP227422SS

 
novus biologicals canada
NBP227422SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1-60 from human BLT1 was used as the immunogen for this antibody (1st extracellular domain).
Antigen Leukotriene B4 R1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human, and white-cheeked gibbon, 94% in common chimpanzee, 83% in orangutan, and 78% in cow, new world monkey, and rhesus monkey.
Gene Accession No. Q15722
Gene ID (Entrez) 1241
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Concentration 0.5mg/mL