Please login to your online account to display your discounted pricing

anti-LDL R, Polyclonal, Novus Biologicals™

$144.63 - $384.00

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the human LDL Receptor protein (within residues 500-550). [Swiss-Prot No. P01130]
Antigen LDL R
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP106709

 
novus biologicals canada
NBP106709
Unlabeled 0.1mL Each for $384.00

NBP106709SS

 
novus biologicals canada
NBP106709SS
Unlabeled 0.025mL Each for $144.63
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the human LDL Receptor protein (within residues 500-550). [Swiss-Prot No. P01130]
Antigen LDL R
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Simple Western
Applications Western Blot
Test Specificity This is specific for both the unglycosylated and glycosylated forms of the LDL Receptor. Human and Mouse. Predicted to react with monkey based on 100% sequence homology.
Gene Accession No. P01130
Gene ID (Entrez) 3949
Formulation PBS, 30% glycerol
Concentration 1.41mg/mL