Please login to your online account to display your discounted pricing

anti-LDL R, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP106709SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP106709SS

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 3949
Formulation PBS, 30% glycerol
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Immunogen Synthetic peptide made to an internal portion of the human LDL Receptor protein (within residues 500-550). [Swiss-Prot No. P01130]
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Simple Western
Applications Western Blotting
Format Affinity Purified
Quantity 0.025mL
Gene Accession No. P01130
Concentration 1.41mg/mL
Test Specificity This is specific for both the unglycosylated and glycosylated forms of the LDL Receptor. Human and Mouse. Predicted to react with monkey based on 100% sequence homology.
Conjugate Unlabeled
Regulatory Status RUO
Cross Reactivity Human
Cross Reactivity Mouse
Host Species Rabbit
Antigen LDL R