Please login to your online account to display your discounted pricing

anti-LC3/MAP1LC3A, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224392SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224392SS

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. Q9H492
Test Specificity The amino acid sequence used as immunogen for the antibody is 100% homologous in human, mouse, rat, chicken, cow, zebrafish and 88% homologous in pufferfish.
Gene ID (Entrez) 84557
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Opossum
Cross Reactivity Rat
Cross Reactivity Zebrafish
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Antigen LC3/MAP1LC3A
Host Species Rabbit
Monoclonal or Polyclonal Polyclonal
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Primary or Secondary Primary
Format Affinity Purified
Immunogen A portion of amino acids 1-50 of human MAP1LC3A was used as the immunogen for the antibody.
Quantity 0.025mg
Regulatory Status RUO