Please login to your online account to display your discounted pricing

anti-LC3/MAP1LC3A, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224392

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224392

  • / Each
 

Description & Specifications

Specifications

Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Gene Accession No. Q9H492
Primary or Secondary Primary
Test Specificity The amino acid sequence used as immunogen for the antibody is 100% homologous in human, mouse, rat, chicken, cow, zebrafish and 88% homologous in pufferfish.
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Opossum
Cross Reactivity Rat
Cross Reactivity Zebrafish
Gene ID (Entrez) 84557
Conjugate Unlabeled
Concentration 0.5mg/mL
Host Species Rabbit
Antigen LC3/MAP1LC3A
Immunogen A portion of amino acids 1-50 of human MAP1LC3A was used as the immunogen for the antibody.
Regulatory Status RUO
Quantity 0.1mg