Please login to your online account to display your discounted pricing

anti-LC3/MAP1LC3A, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP178964SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP178964SS

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 84557
Format Affinity Purified
Concentration 1.09mg/mL
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rat
Antigen LC3/MAP1LC3A
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Quantity 0.025mL
Monoclonal or Polyclonal Polyclonal
Test Specificity This LC3I antibody specifically detects the cytosolic form of LC3 before it gets converted into LC3II during autophagy.
Formulation PBS, 30% glycerol
Gene Accession No. Q9H492
Primary or Secondary Primary
Immunogen A synthetic peptide made to a C-terminal region of the human LC3 protein (within residues 50-120). [Swiss-Prot Q9H492]
Host Species Rabbit
Regulatory Status RUO