Please login to your online account to display your discounted pricing

anti-LC3/MAP1LC3A, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human MAP1LC3A was used as the immunogen for the antibody.
Antigen LC3/MAP1LC3A
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224392

 
novus biologicals canada
NBP224392
Unlabeled 0.1mg Each for $255.80

NBP224392SS

 
novus biologicals canada
NBP224392SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human MAP1LC3A was used as the immunogen for the antibody.
Antigen LC3/MAP1LC3A
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen for the antibody is 100% homologous in human, mouse, rat, chicken, cow, zebrafish and 88% homologous in pufferfish.
Gene Accession No. Q9H492
Gene ID (Entrez) 84557
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL