missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-LC3/MAP1LC3A, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal region of the human LC3 protein (within residues 50-120). [Swiss-Prot Q9H492]
Antigen LC3/MAP1LC3A
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP178964SS Novus Biologicals Canada
NBP178964SS
0.025mL Each for $109.00
NBP178964 Novus Biologicals Canada
NBP178964
0.1mL Each for $309.05
Specifications Description & Specifications

Specifications

Rabbit
LC3/MAP1LC3A
Affinity Purified
RUO
This LC3I antibody specifically detects the cytosolic form of LC3 before it gets converted into LC3II during autophagy.
84557
1.09mg/mL
A synthetic peptide made to a C-terminal region of the human LC3 protein (within residues 50-120). [Swiss-Prot Q9H492]
Polyclonal
Primary
Immunocytochemistry,Immunofluorescence,Immunohistochemistry,Immunohistochemistry (Paraffin),Western Blot
Q9H492
PBS, 30% glycerol