Please login to your online account to display your discounted pricing

anti-Lamin B1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056403SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056403T

  • / Each
 

Description & Specifications

Specifications

Formulation PBS containing 0.05% BSA
Primary or Secondary Primary
Quantity 0.025mg
Regulatory Status RUO
Conjugate Unlabeled
Host Species Rabbit
Gene ID (Entrez) 4001
Applications Western Blotting
Immunogen Full length recombinant Lamin B1 protein was used as the immunogen.
Monoclonal or Polyclonal Polyclonal
Concentration 0.5mg/mL
Cross Reactivity Human
Format Purified
Antigen Lamin B1