Please login to your online account to display your discounted pricing

anti-Lamin B1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056403

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056403

  • / Each
 

Description & Specifications

Specifications

Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL
Conjugate Unlabeled
Primary or Secondary Primary
Antigen Lamin B1
Monoclonal or Polyclonal Polyclonal
Applications Western Blotting
Quantity 0.1mg
Gene ID (Entrez) 4001
Immunogen Full length recombinant Lamin B1 protein was used as the immunogen.
Host Species Rabbit
Format Purified
Regulatory Status RUO
Cross Reactivity Human