Please login to your online account to display your discounted pricing

anti-Lamin B1, Polyclonal, Novus Biologicals™

$139.00 - $274.59

Specifications

Host Species Rabbit
Immunogen Full length recombinant Lamin B1 protein was used as the immunogen.
Antigen Lamin B1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056403T

 
novus biologicals canada
NB10056403SS
Unlabeled 0.025mg Each for $139.00

NB10056403

 
novus biologicals canada
NB10056403
Unlabeled 0.1mg Each for $274.59
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Full length recombinant Lamin B1 protein was used as the immunogen.
Antigen Lamin B1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Gene ID (Entrez) 4001
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL