Please login to your online account to display your discounted pricing

anti-Lamin A + C, Polyclonal, Novus Biologicals™

$109.00 - $307.00

Specifications

Host Species Rabbit
Immunogen Recombinant full length purified Lamin A + C from E. coli
Antigen Lamin A + C
Monoclonal or Polyclonal Polyclonal
Format Antiserum
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP225153

 
novus biologicals canada
NBP225153
Unlabeled 0.1mL Each for $307.00

NBP225153SS

 
novus biologicals canada
NBP225153SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Recombinant full length purified Lamin A + C from E. coli
Antigen Lamin A + C
Monoclonal or Polyclonal Polyclonal
Format Antiserum
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Test Specificity Human, Bovine, Porcine, Mouse, Rat and other Mammals. Other species have not been tested.
Gene Accession No. P02545
Gene ID (Entrez) 4000