Please login to your online account to display your discounted pricing

anti-Lactoperoxidase, Polyclonal, Novus Biologicals™

$109.00 - $300.00

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to a C-terminal portion of the human Lactoperoxidase protein (within residues 650-712). [Swiss-Prot No. P22079]
Antigen Lactoperoxidase
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11061574

 
novus biologicals canada
NB11061574
Unlabeled 0.1mL Each for $300.00

NB11061574T

 
novus biologicals canada
NB11061574SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to a C-terminal portion of the human Lactoperoxidase protein (within residues 650-712). [Swiss-Prot No. P22079]
Antigen Lactoperoxidase
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Gene Accession No. P22079
Gene ID (Entrez) 4025
Formulation PBS, 30% glycerol
Concentration 1.5mg/mL