missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Lactoperoxidase, Polyclonal, Novus Biologicals™

$109.00 - $329.00

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the human Lactoperoxidase protein (within residues 300-400). [Swiss-Prot No. P22079]
Antigen Lactoperoxidase
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB11061572T Novus Biologicals Canada
NB11061572SS
0.025mL Each for $109.00
NB11061572 Novus Biologicals Canada
NB11061572
0.1mL Each for $329.00
Specifications Description & Specifications

Specifications

Rabbit
Lactoperoxidase
Affinity Purified
RUO
Human, monkey and bovine. Immunogen has 81% homology to sheep and 73% homology to mouse and rat.
4025
1.7mg/mL
Synthetic peptide made to an internal portion of the human Lactoperoxidase protein (within residues 300-400). [Swiss-Prot No. P22079]
Polyclonal
Primary
Western Blot
P22079
PBS, 30% glycerol