Please login to your online account to display your discounted pricing

anti-Lactate Dehydrogenase A/LDHA, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to a C-terminal portion of the human Lactate Dehydrogenase A protein (within residues 280-332). [Swiss-Prot No. P00338]
Antigen Lactate Dehydrogenase A/LDHA
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP148336

 
novus biologicals canada
NBP148336
Unlabeled 0.1mL Each for $329.00

NBP148336SS

 
novus biologicals canada
NBP148336SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to a C-terminal portion of the human Lactate Dehydrogenase A protein (within residues 280-332). [Swiss-Prot No. P00338]
Antigen Lactate Dehydrogenase A/LDHA
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blot
Test Specificity Human and mouse. Predicted to react with bovine and porcine based on 100% sequence homology. Bovine data from customer review.
Gene Accession No. P00338
Gene ID (Entrez) 3939
Formulation PBS, 30% glycerol
Concentration 1.25mg/mL