Please login to your online account to display your discounted pricing

anti-Kv1, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to rat Kv1.2 (within residues 50-100)
Antigen Kv1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP142788SS

 
novus biologicals canada
NBP142788SS
Unlabeled 0.025mL Each for $109.00

NBP142788

 
novus biologicals canada
NBP142788
Unlabeled 0.1mL Each for $309.05
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to rat Kv1.2 (within residues 50-100)
Antigen Kv1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunoprecipitation
Applications Western Blot
Test Specificity Reacts with all members of the Kv1 family. Does not cross-react with Kv3.1. Human, mouse, rat, bovine, porcine, canine, Xenopus, Zebrafish and other vertebrates.
Gene ID (Entrez) 3737
Formulation PBS
Concentration 1mg/mL