Please login to your online account to display your discounted pricing

anti-Kv1.2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP142802

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP142802

  • / Each
 

Description & Specifications

Specifications

Formulation PBS
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Regulatory Status RUO
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 3737
Test Specificity Does not cross-react with Kv3.1. Human, mouse, rat, bovine, porcine, canine, Xenopus, Zebrafish and other vertebrates.
Concentration 1mg/mL
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Cross Reactivity Xenopus
Cross Reactivity Zebrafish
Host Species Rabbit
Primary or Secondary Primary
Conjugate Unlabeled
Quantity 0.1mL
Antigen Kv1.2
Format Affinity Purified
Immunogen Synthetic peptide made to a portion of rat Kv1.2 (within residues 50-100). [Swiss-Prot No. P63142]