Please login to your online account to display your discounted pricing

anti-Kv1.2, Polyclonal, Novus Biologicals™

$109.00 - $329.00

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to a portion of rat Kv1.2 (within residues 325-375). [Swiss-Prot No. P63142]
Antigen Kv1.2
Monoclonal or Polyclonal Polyclonal
Format Antiserum
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP142818

 
novus biologicals canada
NBP142818
Unlabeled 0.1mL Each for $329.00

NBP142818SS

 
novus biologicals canada
NBP142818SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to a portion of rat Kv1.2 (within residues 325-375). [Swiss-Prot No. P63142]
Antigen Kv1.2
Monoclonal or Polyclonal Polyclonal
Format Antiserum
Primary or Secondary Primary
Regulatory Status RUO
Applications Blocking
Applications Immunoprecipitation
Applications Neutralization
Applications Western Blotting
Test Specificity Does not cross-react with Kv1.3 and Kv3.1. Human, mouse, rat, rabbit, bovine, porcine, canine, Xenopus and other vertebrates.
Gene ID (Entrez) 3737