Please login to your online account to display your discounted pricing

anti-Kv1.2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP142818SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP142818SS

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3737
Primary or Secondary Primary
Quantity 0.025mL
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rabbit
Cross Reactivity Rat
Cross Reactivity Xenopus
Immunogen Synthetic peptide made to a portion of rat Kv1.2 (within residues 325-375). [Swiss-Prot No. P63142]
Applications Blocking
Applications Immunoprecipitation
Applications Neutralization
Applications Western Blotting
Test Specificity Does not cross-react with Kv1.3 and Kv3.1. Human, mouse, rat, rabbit, bovine, porcine, canine, Xenopus and other vertebrates.
Monoclonal or Polyclonal Polyclonal
Format Antiserum
Regulatory Status RUO
Host Species Rabbit
Antigen Kv1.2