Please login to your online account to display your discounted pricing

anti-Kv1.2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP142818

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP142818

  • / Each
 

Description & Specifications

Specifications

Antigen Kv1.2
Applications Blocking
Applications Immunoprecipitation
Applications Neutralization
Applications Western Blot
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rabbit
Cross Reactivity Rat
Cross Reactivity Xenopus
Format Antisera
Host Species Rabbit
Immunogen Synthetic peptide made to a portion of rat Kv1.2 (within residues 325-375). [Swiss-Prot No. P63142]
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 3737
Test Specificity Does not cross-react with Kv1.3 and Kv3.1. Human, mouse, rat, rabbit, bovine, porcine, canine, Xenopus and other vertebrates.
Target Species Bovine
Target Species Canine
Target Species Human
Target Species Murine
Target Species Porcine
Target Species Rabbit
Target Species Rat
Target Species Xenopus