Please login to your online account to display your discounted pricing

anti-Kv1.2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP142802SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP142802SS

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 3737
Formulation PBS
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Test Specificity Does not cross-react with Kv3.1. Human, mouse, rat, bovine, porcine, canine, Xenopus, Zebrafish and other vertebrates.
Antigen Kv1.2
Regulatory Status RUO
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Cross Reactivity Xenopus
Cross Reactivity Zebrafish
Format Affinity Purified
Host Species Rabbit
Immunogen Synthetic peptide made to a portion of rat Kv1.2 (within residues 50-100). [Swiss-Prot No. P63142]
Quantity 0.025mL