Please login to your online account to display your discounted pricing

anti-KT3 Epitope Tag, Polyclonal, Novus Biologicals™

$274.59 - $993.54

Specifications

Host Species Rabbit
Immunogen Rabbits were immunized with KT3 (KPPTPPPEPET) conjugated to KLH. Antibody was isolated by affinity chromatography using the peptide immobilized on solid support.
Antigen KT3 Epitope Tag
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11040655

 
novus biologicals canada
NB11040655
Unlabeled 0.1mg Each for $274.59

NB110406551

 
novus biologicals canada
NB110406551MG
Unlabeled 1mg Each for $993.54
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Rabbits were immunized with KT3 (KPPTPPPEPET) conjugated to KLH. Antibody was isolated by affinity chromatography using the peptide immobilized on solid support.
Antigen KT3 Epitope Tag
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Formulation phosphate Buffered Saline (PBS)
Concentration 1.0mg/mL