Please login to your online account to display your discounted pricing

anti-KT3 Epitope Tag, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB110406551MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB110406551

  • / Each
 

Description & Specifications

Specifications

Formulation phosphate Buffered Saline (PBS)
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Concentration 1.0mg/mL
Conjugate Unlabeled
Host Species Rabbit
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Regulatory Status RUO
Cross Reactivity Non-species specific
Immunogen Rabbits were immunized with KT3 (KPPTPPPEPET) conjugated to KLH. Antibody was isolated by affinity chromatography using the peptide immobilized on solid support.
Antigen KT3 Epitope Tag
Format Affinity Purified
Quantity 1mg