Please login to your online account to display your discounted pricing

anti-KLF8, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1-50 from human kLF8 was used as the immunogen for this antibody.
Antigen KLF8
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227418SS

 
novus biologicals canada
NBP227418SS
Unlabeled 0.025mg Each for $109.00

NBP227418

 
novus biologicals canada
NBP227418
Unlabeled 0.1mg Each for $255.80
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1-50 from human kLF8 was used as the immunogen for this antibody.
Antigen KLF8
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human, sumatran orangutan, and white-cheeked gibbon, 89% in rhesus monkey, and 78% in new world monkey.
Gene Accession No. O95600
Gene ID (Entrez) 11279
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Concentration 0.5mg/mL