Please login to your online account to display your discounted pricing

anti-KIR3DL2/CD158k, PerCP, Clone: 539304, R&D Systems


Manufacturer: r & d systems  RDSFAB2878C025

 View more versions of this product

Catalog No. FAB2878C025

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Format Purified
Cross Reactivity Human
Regulatory Status RUO
Antigen KIR3DL2/CD158k
Conjugate PerCP
Primary or Secondary Primary
Immunogen BaF3 mouse pro-B cell line transfected with human KIR3DL2/CD158k.
Clone 539304
Isotype IgG1
Quantity 25 Tests
Purification Method Protein A or G purified from hybridoma culture supernatant
Monoclonal or Polyclonal Monoclonal