Please login to your online account to display your discounted pricing

anti-KIR3DL2/CD158k, PerCP, Clone: 539304, R&D Systems

$142.08 - $428.87

Specifications

Antigen KIR3DL2/CD158k
Clone 539304
Cross Reactivity Human
Format Purified
Host Species Mouse
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

FAB2878C025

 
r & d systems
RDSFAB2878C025
PerCP 25 Tests Each for $142.08

FAB2878C100

 
r & d systems
RDSFAB2878C100
PerCP 100 Tests Each for $428.87
Description & Specifications

Specifications

Antigen KIR3DL2/CD158k
Clone 539304
Cross Reactivity Human
Format Purified
Host Species Mouse
Regulatory Status RUO
Immunogen BaF3 mouse pro-B cell line transfected with human KIR3DL2/CD158k.
Isotype IgG1
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Purification Method Protein A or G purified from hybridoma culture supernatant

Applications:

Flow Cytometry