Please login to your online account to display your discounted pricing

anti-KIR3DL2/CD158k, PerCP, Clone: 539304, R&D Systems


Manufacturer: r & d systems  RDSFAB2878C100

 View more versions of this product

Catalog No. FAB2878C100

  • / Each
 

Description & Specifications

Specifications

Antigen KIR3DL2/CD158k
Format Purified
Host Species Mouse
Immunogen BaF3 mouse pro-B cell line transfected with human KIR3DL2/CD158k.
Isotype IgG1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone 539304
Conjugate PerCP
Cross Reactivity Human