Please login to your online account to display your discounted pricing

anti-KIR3DL2/CD158k, PE, Clone: 539304, R&D Systems


Manufacturer: r & d systems  RDSFAB2878P025

 View more versions of this product

Catalog No. FAB2878P025

  • / Each
 

Description & Specifications

Specifications

Isotype IgG1
Antigen KIR3DL2/CD158k
Immunogen BaF3 mouse pro-B cell line transfected with human KIR3DL2/CD158k.
Format Purified
Regulatory Status RUO
Cross Reactivity Human
Quantity 25 Tests
Primary or Secondary Primary
Conjugate PE
Monoclonal or Polyclonal Monoclonal
Host Species Mouse
Clone 539304
Purification Method Protein A or G purified from hybridoma culture supernatant