Please login to your online account to display your discounted pricing

anti-KIR3DL2/CD158k, PE, Clone: 539304, R&D Systems


Manufacturer: r & d systems  RDSFAB2878P100

 View more versions of this product

Catalog No. FAB2878P100

  • / Each
 

Description & Specifications

Specifications

Format Purified
Isotype IgG1
Quantity 100 Tests
Antigen KIR3DL2/CD158k
Host Species Mouse
Immunogen BaF3 mouse pro-B cell line transfected with human KIR3DL2/CD158k.
Purification Method Protein A or G purified from hybridoma culture supernatant
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone 539304
Conjugate PE
Cross Reactivity Human