Please login to your online account to display your discounted pricing

anti-KIR3DL1, PE, Clone: DX9, R&D Systems


Manufacturer: r & d systems  RDSFAB1225P025

 View more versions of this product

Catalog No. FAB1225P025

  • / Each
 

Description & Specifications

Specifications

Format Purified
Host Species Mouse
Immunogen Human KIR3DL1-expressing NK cell clone.
Isotype IgG1
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Conjugate PE
Cross Reactivity Human
Antigen KIR3DL1
Purification Method Protein A or G purified from hybridoma culture supernatant
Clone DX9