Please login to your online account to display your discounted pricing

anti-KIR3DL1, PE, Clone: DX9, R&D Systems

$126.32 - $394.29

Specifications

Antigen KIR3DL1
Clone DX9
Cross Reactivity Human
Format Purified
Host Species Mouse
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

FAB1225P025

 
r & d systems
RDSFAB1225P025
PE 25 Tests Each for $126.32

FAB1225P100

 
r & d systems
RDSFAB1225P100
PE 100 Tests Each for $394.29
Description & Specifications

Specifications

Antigen KIR3DL1
Clone DX9
Cross Reactivity Human
Format Purified
Host Species Mouse
Regulatory Status RUO
Immunogen Human KIR3DL1-expressing NK cell clone.
Isotype IgG1
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Purification Method Protein A or G purified from hybridoma culture supernatant

Applications:

Flow Cytometry