Please login to your online account to display your discounted pricing

anti-KIR3DL1, APC, Clone: DX9, R&D Systems


Manufacturer: r & d systems  RDSFAB1225A025

 View more versions of this product

Catalog No. FAB1225A025

  • / Each
 

Description & Specifications

Specifications

Antigen KIR3DL1
Clone DX9
Conjugate Allophycocyanin
Cross Reactivity Human
Format Purified
Host Species Murine
Immunogen Human KIR3DL1-expressing NK cell clone.
Isotype IgG1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Target Species Human