Please login to your online account to display your discounted pricing

anti-KIR2DS4/CD158i, PE, Clone: 179315, R&D Systems


Manufacturer: r & d systems  RDSFAB1847P100

 View more versions of this product

Catalog No. FAB1847P100

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Cross Reactivity Human
Antigen KIR2DS4/CD158i
Quantity 100 Tests
Isotype IgG2a
Clone 179315
Conjugate PE
Format Purified
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Regulatory Status RUO
Immunogen BaF3 mouse pro-B cell line transfected with human KIR2DS4.
Purification Method Protein A or G purified from hybridoma culture supernatant