Please login to your online account to display your discounted pricing

anti-KIR2DS4/CD158i, PE, Clone: 179315, R&D Systems


Manufacturer: r & d systems  RDSFAB1847P025

 View more versions of this product

Catalog No. FAB1847P025

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Quantity 25 Tests
Antigen KIR2DS4/CD158i
Immunogen BaF3 mouse pro-B cell line transfected with human KIR2DS4.
Regulatory Status RUO
Clone 179315
Conjugate PE
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Isotype IgG2a
Format Purified
Host Species Mouse
Purification Method Protein A or G purified from hybridoma culture supernatant