Please login to your online account to display your discounted pricing

anti-KIR2DS4/CD158i, PE, Clone: 179315, R&D Systems

$129.05 - $337.62

Specifications

Antigen KIR2DS4/CD158i
Clone 179315
Cross Reactivity Human
Format Purified
Host Species Mouse
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

FAB1847P025

 
r & d systems
RDSFAB1847P025
PE 25 Tests Each for $129.05

FAB1847P100

 
r & d systems
RDSFAB1847P100
PE 100 Tests Each for $337.62
Description & Specifications

Specifications

Antigen KIR2DS4/CD158i
Clone 179315
Cross Reactivity Human
Format Purified
Host Species Mouse
Regulatory Status RUO
Immunogen BaF3 mouse pro-B cell line transfected with human KIR2DS4.
Isotype IgG2a
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Purification Method Protein A or G purified from hybridoma culture supernatant

Applications:

Flow Cytometry