Please login to your online account to display your discounted pricing

anti-KIR2DS4/CD158i, APC, Clone: 179315, R&D Systems


Manufacturer: r & d systems  RDSFAB1847A100

 View more versions of this product

Catalog No. FAB1847A100

  • / Each
 

Description & Specifications

Specifications

Format Purified
Immunogen BaF3 mouse pro-B cell line transfected with human KIR2DS4.
Cross Reactivity Human
Quantity 100 Tests
Regulatory Status RUO
Host Species Mouse
Antigen KIR2DS4/CD158i
Primary or Secondary Primary
Conjugate APC
Monoclonal or Polyclonal Monoclonal
Clone 179315
Isotype IgG2a
Purification Method Protein A or G purified from hybridoma culture supernatant