Please login to your online account to display your discounted pricing

anti-KIR2DS4/CD158i, APC, Clone: 179315, R&D Systems


Manufacturer: r & d systems  RDSFAB1847A025

 View more versions of this product

Catalog No. FAB1847A025

  • / Each
 

Description & Specifications

Specifications

Antigen KIR2DS4/CD158i
Format Purified
Cross Reactivity Human
Isotype IgG2a
Quantity 25 Tests
Clone 179315
Host Species Mouse
Immunogen BaF3 mouse pro-B cell line transfected with human KIR2DS4.
Purification Method Protein A or G purified from hybridoma culture supernatant
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Conjugate APC