Please login to your online account to display your discounted pricing

anti-KIR2DL4/CD158d, PE, Clone: 181703, R&D Systems

$142.08 - $363.69

Specifications

Antigen KIR2DL4/CD158d
Clone 181703
Cross Reactivity Human
Format Purified
Host Species Mouse
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

FAB2238P025

 
r & d systems
RDSFAB2238P025
PE 25 Tests Each for $142.08

FAB2238P100

 
r & d systems
RDSFAB2238P100
PE 100 Tests Each for $363.69
Description & Specifications

Specifications

Antigen KIR2DL4/CD158d
Clone 181703
Cross Reactivity Human
Format Purified
Host Species Mouse
Regulatory Status RUO
Immunogen BaF3 mouse pro-B cell line transfected with human KIR2DL4.
Isotype IgG2a
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Purification Method Protein A or G purified from hybridoma culture supernatant

Applications:

Flow Cytometry