Please login to your online account to display your discounted pricing

anti-KIR2DL4/CD158d, Fluorescein, Clone: 181703, R&D Systems


Manufacturer: r & d systems  RDSFAB2238F100

 View more versions of this product

Catalog No. FAB2238F100

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Isotype IgG2a
Quantity 100 Tests
Cross Reactivity Human
Format Purified
Regulatory Status RUO
Clone 181703
Antigen KIR2DL4/CD158d
Immunogen BaF3 mouse pro-B cell line transfected with human KIR2DL4.
Purification Method Protein A or G purified from hybridoma culture supernatant
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Conjugate Fluorescein