Please login to your online account to display your discounted pricing

anti-KIR2DL4/CD158d, APC, Clone: 181703, R&D Systems

$139.08 - $407.05

Specifications

Antigen KIR2DL4/CD158d
Clone 181703
Cross Reactivity Human
Format Purified
Host Species Mouse
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

FAB2238A025

 
r & d systems
RDSFAB2238A025
APC 25 Tests Each for $139.08

FAB2238A100

 
r & d systems
RDSFAB2238A100
APC 100 Tests Each for $407.05
Description & Specifications

Specifications

Antigen KIR2DL4/CD158d
Clone 181703
Cross Reactivity Human
Format Purified
Host Species Mouse
Regulatory Status RUO
Immunogen BaF3 mouse pro-B cell line transfected with human KIR2DL4.
Isotype IgG2a
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Purification Method Protein A or G purified from hybridoma culture supernatant

Applications:

Flow Cytometry