Please login to your online account to display your discounted pricing

anti-KIR2DL3/CD158b2, PerCP, Clone: 180701, R&D Systems


Manufacturer: r & d systems  RDSFAB2014C025

 View more versions of this product

Catalog No. FAB2014C025

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Clone 180701
Regulatory Status RUO
Format Purified
Isotype IgG2a
Immunogen BaF3 mouse pro-B cell line transfected with human KIR2DL3/CD158b2.
Cross Reactivity Human
Primary or Secondary Primary
Conjugate PerCP
Monoclonal or Polyclonal Monoclonal
Quantity 25 Tests
Antigen KIR2DL3/CD158b2
Purification Method Protein A or G purified from hybridoma culture supernatant