Please login to your online account to display your discounted pricing

anti-KIR2DL3/CD158b2, PerCP, Clone: 180701, R&D Systems

$164.61 - $419.81

Specifications

Antigen KIR2DL3/CD158b2
Clone 180701
Cross Reactivity Human
Format Purified
Host Species Mouse
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

FAB2014C025

 
r & d systems
RDSFAB2014C025
PerCP 25 Tests Each for $164.61

FAB2014C100

 
r & d systems
RDSFAB2014C100
PerCP 100 Tests Each for $419.81
Description & Specifications

Specifications

Antigen KIR2DL3/CD158b2
Clone 180701
Cross Reactivity Human
Format Purified
Host Species Mouse
Regulatory Status RUO
Immunogen BaF3 mouse pro-B cell line transfected with human KIR2DL3/CD158b2.
Isotype IgG2a
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Purification Method Protein A or G purified from hybridoma culture supernatant

Applications:

Flow Cytometry